3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester, 95%
3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester, 95%
3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester, 95%
Thermo Scientific Chemicals

3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester, 95%

CAS: 1408088-34-5 | C17H26BFN2O2 | 320.215 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H59640.03
also known as H59640-03
Price (USD)
299.65
Online Exclusive
333.00
Save 33.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
299.65
Online Exclusive
333.00
Save 33.35 (10%)
Each
Specifications
CAS1408088-34-5
Chemical Name or Material3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester
Formula Weight320.21
IUPAC Name1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4-methylpiperazine
InChI KeyRFECQDXNBWGCRR-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only