Methyl hydrogen succinate, 95%
Methyl hydrogen succinate, 95%
Methyl hydrogen succinate, 95%
Thermo Scientific Chemicals

Methyl hydrogen succinate, 95%

CAS: 3878-55-5 | C5H8O4 | 132.115 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H59722.14
also known as H59722-14
Price (USD)
81.65
Online exclusive
90.50
Save 8.85
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
81.65
Online exclusive
90.50
Save 8.85
Each
Chemical Identifiers
CAS3878-55-5
IUPAC Name4-methoxy-4-oxobutanoic acid
Molecular FormulaC5H8O4
InChI KeyJDRMYOQETPMYQX-UHFFFAOYSA-N
SMILESCOC(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥94.0%
Appearance (Color)White
FormPowder or crystals or crystalline powder
Melting Point56.0-61.0°C
It could react with 1,2,3-trimethoxy-benzene, and obtain the 4-oxo-4-(2,3,4-trimethoxy-phenyl)-butyric acid methyl ester. This reaction needs the reagent of PPA. Butanedioic Acid 1-Methyl Ester is an intermediate in the synthesis of agonist to both GHB and GABAB neural receptors, exhibiting neurotransmitter-like effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It could react with 1,2,3-trimethoxy-benzene, and obtain the 4-oxo-4-(2,3,4-trimethoxy-phenyl)-butyric acid methyl ester. This reaction needs the reagent of PPA. Butanedioic Acid 1-Methyl Ester is an intermediate in the synthesis of agonist to both GHB and GABAB neural receptors, exhibiting neurotransmitter-like effects.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Ryszard Andruszkiewicz, et al. Synthesis of N3-fumaramoyl-L-2,3-diaminopropanoic acid analogues, the irreversible inhibitors of glucosamine synthetase.Int. J. Pept. Protein Res.,1986,27(5), 449-453.
  2. P Borgeat, et al. Transformation of arachidonic acid and homo-gamma-linolenic acid by rabbit polymorphonuclear leukocytes. Monohydroxy acids from novel lipoxygenases.J. Biol. Chem.,1976,2527816-7820.