Quinoxaline-2-carboxylic acid, 97%
Quinoxaline-2-carboxylic acid, 97%
Quinoxaline-2-carboxylic acid, 97%
Thermo Scientific Chemicals

Quinoxaline-2-carboxylic acid, 97%

CAS: 879-65-2 | C9H6N2O2 | 174.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number H59780.ME
also known as H59780-ME
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Quantity:
500 mg
Request bulk or custom format
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Chemical Identifiers
CAS879-65-2
IUPAC Namequinoxaline-2-carboxylic acid
Molecular FormulaC9H6N2O2
InChI KeyUPUZGXILYFKSGE-UHFFFAOYSA-N
SMILESOC(=O)C1=CN=C2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only