L-Aspartic acid 4-tert-butyl ester, 98%
L-Aspartic acid 4-tert-butyl ester, 98%
L-Aspartic acid 4-tert-butyl ester, 98%
L-Aspartic acid 4-tert-butyl ester, 98%
Thermo Scientific Chemicals

L-Aspartic acid 4-tert-butyl ester, 98%

CAS: 3057-74-7 | C8H15NO4 | 189.211 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H59810.03
also known as H59810-03
Price (USD)
88.65
Online Exclusive
98.40
Save 9.75 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
88.65
Online Exclusive
98.40
Save 9.75 (10%)
Each
Add to cart
Chemical Identifiers
CAS3057-74-7
IUPAC Name2-amino-4-(tert-butoxy)-4-oxobutanoic acid
Molecular FormulaC8H15NO4
InChI KeyMXWMFBYWXMXRPD-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)CC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only