4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 96%
4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 96%
4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 96%
Thermo Scientific Chemicals

4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 96%

CAS: 1029439-49-3 | C13H17BBrFO2 | 314.989 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H59821.MD
also known as H59821-MD
Price (USD)
105.65
Online Exclusive
117.00
Save 11.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
105.65
Online Exclusive
117.00
Save 11.35 (10%)
Each
Specifications
CAS1029439-49-3
Chemical Name or Material4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester
Formula Weight314.99
IUPAC Name2-[4-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
InChI KeyQOCYLABQSVWXGE-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only