N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals
N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals
N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals
N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H59838.06
also known as H59838-06
5 g
Catalog number H59838.06
also known as H59838-06
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS5545-52-8
IUPAC Name(2S)-2-{[(benzyloxy)carbonyl]amino}-4-(tert-butoxy)-4-oxobutanoic acid
Molecular FormulaC16H21NO6
InChI KeyHLSLRFBLVZUVIE-LBPRGKRZSA-N
SMILESCC(C)(C)OC(=O)C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Solubility
Soluble in DMSO, and methanol.

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. William Spillane.; Jean-Baptiste Malaubier. Sulfamic Acid and Its N- and O-Substituted Derivatives. Chem. Rev. 2014, 114 (4),2507-2586.
  2. Jorge Heredia-Moya.; Kenneth L. Kirk. Synthesis of β-(S-methyl)thioaspartic acid and derivatives. Bioorg. Med. Chem. 2008, 16 (11),5908-5913.