3-Methyl-1H-pyrazole-5-carboxylic acid, 97%
3-Methyl-1H-pyrazole-5-carboxylic acid, 97%
3-Methyl-1H-pyrazole-5-carboxylic acid, 97%
Thermo Scientific Chemicals

3-Methyl-1H-pyrazole-5-carboxylic acid, 97%

CAS: 402-61-9 | C5H6N2O2 | 126.115 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H59899.06
also known as H59899-06
Price (USD)
398.65
Online Exclusive
443.00
Save 44.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
398.65
Online Exclusive
443.00
Save 44.35 (10%)
Each
Chemical Identifiers
CAS402-61-9
IUPAC Name5-methyl-1H-pyrazole-3-carboxylic acid
Molecular FormulaC5H6N2O2
InChI KeyWSMQKESQZFQMFW-UHFFFAOYSA-N
SMILESCC1=CC(=NN1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
3-Methylpyrazole-5-carboxylic acid is a potent and selective D-amino acid oxidase (DAO) (also known as DAAO, DAMOX and OXDA) inhibitor that protects DAO cells from oxidative stress induced by D-Serine. 3-Methylpyrazole-5-carboxylic acid specifically prevents formalin-induced tonic pain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylpyrazole-5-carboxylic acid is a potent and selective D-amino acid oxidase (DAO) (also known as DAAO, DAMOX and OXDA) inhibitor that protects DAO cells from oxidative stress induced by D-Serine. 3-Methylpyrazole-5-carboxylic acid specifically prevents formalin-induced tonic pain.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Donald J. P. Pinto.; Michael J. Orwat.; Shuaige Wang.; John M. Fevig.; Mimi L. Quan.; Eugene Amparo.; Joseph Cacciola.; Karen A. Rossi.; Richard S. Alexander.; Angela M. Smallwood.; Joseph M. Luettgen.; Li Liang.; Bruce J. Aungst.; Matthew R. Wright.; Robert M. Knabb.; Pancras C. Wong.; Ruth R. Wexler.; Patrick Y. S. Lam. Discovery of 1-[3-(Aminomethyl)phenyl]-N-[3-fluoro-2‘-(methylsulfonyl)- [1,1‘-biphenyl]-4-yl]-3-(trifluoromethyl)-1H-pyrazole-5-carboxamide (DPC423), a Highly Potent, Selective, and Orally Bioavailable Inhibitor of Blood Coagulation Factor Xa1.J. Med. Chem. 2001, 44 (4), 566-578.
  2. James R. Pruitt.; Donald J. P. Pinto.; Robert A. Galemmo, Jr.; Richard S. Alexander.; Karen A. Rossi.; Brian L. Wells.; Spencer Drummond.; Lori L. Bostrom.; Debra Burdick.; Robert Bruckner.; Haiying Chen.; Angela Smallwood.; Pancras C. Wong.; Matthew R. Wright.; Steven Bai.; Joseph M. Luettgen. ;Robert M. Knabb.; Patrick Y. S. Lam.; Ruth R. Wexler. Discovery of 1-(2-Aminomethylphenyl)-3-trifluoromethyl-N- [3-fluoro-2‘-(aminosulfonyl)[1,1‘-biphenyl)]-4-yl]-1H-pyrazole-5-carboxyamide (DPC602), a Potent, Selective, and Orally Bioavailable Factor Xa Inhibitor1. J. Med. Chem. 2003, 46 (25),5298-5315 .