4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H59944.03
also known as H59944-03
1 g
Catalog number H59944.03
also known as H59944-03
Price (USD)
322.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
322.00
Each
Specifications
CAS1326283-60-6
Chemical Name or Material4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester
Formula Weight267.11
IUPAC Name5-fluoro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
InChI KeyJWACNMHKJHEWAQ-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only