1-Benzyl-3-methylimidazolium chloride, 97%
1-Benzyl-3-methylimidazolium chloride, 97%
1-Benzyl-3-methylimidazolium chloride, 97%
Thermo Scientific Chemicals

1-Benzyl-3-methylimidazolium chloride, 97%

CAS: 36443-80-8 | C11H13ClN2 | 208.689 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number H59997.06
also known as H59997-06
Price (USD)
39.20
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.20
Each
Chemical Identifiers
CAS36443-80-8
IUPAC Name1-benzyl-3-methyl-1H-imidazol-3-ium chloride
Molecular FormulaC11H13ClN2
InChI KeyFCRZSGZCOGHOGF-UHFFFAOYSA-M
SMILES[Cl-].C[N+]1=CN(CC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only