Oxcarbazepine, 98%
Oxcarbazepine, 98%
Oxcarbazepine, 98%
Thermo Scientific Chemicals

Oxcarbazepine, 98%

CAS: 28721-07-5 | C15H12N2O2 | 252.273 g/mol
Have Questions?
Catalog NumberQuantity
H60014.MD
also known as H60014-MD
250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H60014.MD
also known as H60014-MD
Price (USD)
158.65
Online Exclusive
176.00
Save 17.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
158.65
Online Exclusive
176.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS28721-07-5
IUPAC Name9-oxo-2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene-2-carboxamide
Molecular FormulaC15H12N2O2
InChI KeyCTRLABGOLIVAIY-UHFFFAOYSA-N
SMILESNC(=O)N1C2=CC=CC=C2CC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
Oxcarbazepine is a sodium channel protein inhibitor. It is an anticonvulsant and mood-stab.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in DMSO, methanol, water, ethanol and acetone.

Notes
Store at -20°C.
RUO – Research Use Only

General References:

  1. Mogens Dam (Chairman).; Rolf Ekberg.; Yngve Løyning.; Olli Waltimo.; Kurt Jakobsen. A double-blind study comparing oxcarbazepine and carbamazepine in patients with newly diagnosed, previously untreated epilepsy. Epilepsy Research. 1989, 3 (1),70-76.
  2. M. L. Friis.;O. Kristensen.; J. Boas.; M. Dalby.; S. H. Deth.; L. Gram.; M. Mikkelsen.; B. Pedersen.; A. Sabers.; J. Worm-Petersen.; D. Andersen.; P. K. Jensen. Therapeutic experiences with 947 epileptic out-patients in oxcarbazepine treatment. Acta Neurologica Scandinavica. 1993, 87 (3),224-227.