2-Bromo-1-methylbenzimidazole, 97%
2-Bromo-1-methylbenzimidazole, 97%
2-Bromo-1-methylbenzimidazole, 97%
Thermo Scientific Chemicals

2-Bromo-1-methylbenzimidazole, 97%

CAS: 49572-60-3 | C8H7BrN2 | 211.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H60075.06
also known as H60075-06
Price (USD)
763.65
Online Exclusive
848.00
Save 84.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
763.65
Online Exclusive
848.00
Save 84.35 (10%)
Each
Specifications
CAS49572-60-3
Chemical Name or Material2-Bromo-1-methylbenzimidazole
Formula Weight211.06
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
It is a synthetic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a synthetic building block.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Fei Wang.; Shangjun Cai.; Qian Liao. ;Chanjuan Xi. A Protocol to 2-Aminobenzimidazoles via Copper-Catalyzed Cascade Addition and Cyclization of o-Haloanilines and Carbodiimides. J. Org. Chem. 2011, 76 (9),3174-3180.
  2. Heather M. Lima.; Carl J. Lovely. Synthesis of 2-Imidazolones and 2-Iminoimidazoles. Org. Lett. 2011, 13 (21),5736-5739.