Adamantane-1,3-dicarboxylic acid, 98%
Adamantane-1,3-dicarboxylic acid, 98%
Adamantane-1,3-dicarboxylic acid, 98%
Adamantane-1,3-dicarboxylic acid, 98%
Thermo Scientific Chemicals

Adamantane-1,3-dicarboxylic acid, 98%

CAS: 39269-10-8 | C12H14O4 | 222.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H60081.03
also known as H60081-03
Price (USD)
56.65
Online Exclusive
62.70
Save 6.05 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
62.70
Save 6.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS39269-10-8
IUPAC Nameadamantane-1,3-dicarboxylate
Molecular FormulaC12H14O4
InChI KeyPAVQGHWQOQZQEH-UHFFFAOYSA-L
SMILES[O-]C(=O)C12CC3CC(C1)CC(C3)(C2)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only