(1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine, 97+%
(1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine, 97+%
(1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine, 97+%
(1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine, 97+%
Thermo Scientific Chemicals

(1R,2R)-(-)-2-(Diphenylphosphino)cyclohexylamine, 97+%

CAS: 452304-59-5 | C18H22NP | 283.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H60084.03
also known as H60084-03
Price (USD)
435.00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
435.00
Each
Add to cart
Chemical Identifiers
CAS452304-59-5
IUPAC Name(1R,2R)-2-(diphenylphosphanyl)cyclohexan-1-amine
Molecular FormulaC18H22NP
InChI KeyZATLZEHZPXYMFE-YTXKPMTANA-N
SMILESN[C@@H]1CCCC[C@H]1P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow to pale brown
FormPowder or crystalline powder or crystals
Assay by NMR≥97.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Insoluble in water.

Notes
Air sensitive. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Osamu Kitagawa.; Kanako Yotsumoto.; Mitsuteru Kohriyama.; Yasuo Dobashi.; Takeo Taguchi. Catalytic Asymmetric Synthesis of Vicinal Diamine Derivatives through Enantioselective N-Allylation Using Chiral π-Allyl Pd-Catalyst.Org. Lett. 2004, 6 (20),3605-3607 .