(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%
(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%
(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%
Thermo Scientific Chemicals

(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%

CAS: 1365531-98-1 | C76H108O8P2 | 1211.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H60112.03
also known as H60112-03
Price (USD)
704.65
Online Exclusive
783.00
Save 78.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
704.65
Online Exclusive
783.00
Save 78.35 (10%)
Each
Chemical Identifiers
CAS1365531-98-1
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97%
Enantiomeric excessChiral HPLC or by 1H nmr
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Insoluble in water.

Notes
Incompatible with light and oxidizing agents. Store in a cool place.
RUO – Research Use Only
Ch Durga Prasad.; Shah Jaimin Balkrishna.; Amit Kumar.; Bhagat Singh Bhakuni.; Kaustubh Shrimali.; Soumava Biswas.; Sangit Kumar. Transition-Metal-Free Synthesis of Unsymmetrical Diaryl Chalcogenides from Arenes and Diaryl Dichalcogenides. J. Org. Chem. 2013, 78 (4), 1434-1443.