kappa-Carrageenan
kappa-Carrageenan
kappa-Carrageenan
Thermo Scientific Chemicals

kappa-Carrageenan

CAS: 11114-20-8 | C24H36O25S2-2 | 788.647 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H60117.06
also known as H60117-06
Price (USD)
51.90
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
51.90
Each
Add to cart
Chemical Identifiers
CAS11114-20-8
IUPAC Name(2R,3R,4R,5R,6S)-6-{[(1R,3R,4R,5R,8S)-3-{[(2S,3R,4R,5S,6R)-2-{[(1R,3S,4R,5R,8S)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3-hydroxy-6-(hydroxymethyl)-5-(sulfonatooxy)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl sulfate
Molecular FormulaC24H36O25S2
InChI KeyZNOZWUKQPJXOIG-XSBHQQIPSA-L
SMILESOC[C@H]1O[C@@H](O[C@H]2[C@H]3CO[C@@H]2[C@@H](O)[C@@H](O[C@@H]2[C@@H](O)[C@H](O[C@H]4[C@H]5CO[C@@H]4[C@@H](O)[C@@H](O)O5)O[C@H](CO)[C@@H]2OS([O-])(=O)=O)O3)[C@H](O)[C@@H](O)[C@H]1OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Loss on Drying≤12%
Appearance (Color)White to cream to orange to pale brown to brown or gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only