Specifications
CAS3381-73-5
Chemical Name or Materialalpha-Bromo-4-chlorophenylacetic acid
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Formula Weight249.49
Health Hazard 1H314-H335
View more