Terpineol, mixed isomers, 98%
Terpineol, mixed isomers, 98%
Terpineol, mixed isomers, 98%
Thermo Scientific Chemicals

Terpineol, mixed isomers, 98%

CAS: 8000-41-7 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Catalog number H60146.K4
also known as H60146-K4
Price (USD)
418.00
Each
Quantity:
2.5 L
Request bulk or custom format
Price (USD)
418.00
Each
Chemical Identifiers
CAS8000-41-7
IUPAC Name2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
Molecular FormulaC10H18O
InChI KeyWUOACPNHFRMFPN-UHFFFAOYNA-N
SMILESCC1=CCC(CC1)C(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormLiquid or solidified melt
Assay (GC)>97.5%
Terpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Terpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.

Solubility
Miscible with organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Prakash, B.; Singh, P.; Goni, R.; Raina, A. K.; Dubey, N. K. Efficacy of essential oil, phenyl ethyl alcohol and alfa-terpineol against isolated molds from walnut and their antiaflatoxigenic and antioxidant activity. J. Food Sci. Technol. 2015, 52 (4), 2220-2228.
  2. Lee, S. Y.; Kim, S. H.; Hong, C. Y.; Park, S. Y.; Choi, I. G. Biotransformation of (-)-α-pinene and geraniol to alfa-terpineol and p-menthane-3, 8-diol by the white rot fungus, Polyporus brumalis. J. Microbiol. 2015, 53 (7), 462-467.