(-)-beta-Citronellol, 97%
(-)-beta-Citronellol, 97%
(-)-beta-Citronellol, 97%
(-)-beta-Citronellol, 97%
Thermo Scientific Chemicals

(-)-beta-Citronellol, 97%

CAS: 7540-51-4 | C10H20O | 156.269 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H60174.06
also known as H60174-06
Price (USD)
122.65
Special offer
Online exclusive
Ends: 30-Jun-2026
144.00
Save 21.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
122.65
Special offer
Online exclusive
Ends: 30-Jun-2026
144.00
Save 21.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7540-51-4
IUPAC Name(3S)-3,7-dimethyloct-6-en-1-ol
Molecular FormulaC10H20O
InChI KeyQMVPMAAFGQKVCJ-JTQLQIEISA-N
SMILESC[C@H](CCO)CCC=C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only