Folinic acid calcium salt hydrate, 98%
Folinic acid calcium salt hydrate, 98%
Folinic acid calcium salt hydrate, 98%
Thermo Scientific Chemicals

Folinic acid calcium salt hydrate, 98%

CAS: 1492-18-8 | C20H21CaN7O7 | 511.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H60181.03
also known as H60181-03
Price (USD)
421.65
Online Exclusive
468.00
Save 46.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
421.65
Online Exclusive
468.00
Save 46.35 (10%)
Each
Chemical Identifiers
CAS1492-18-8
IUPAC Namecalcium (2S)-2-[(4-{[(2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioate
Molecular FormulaC20H21CaN7O7
InChI KeyKVUAALJSMIVURS-AEQORCIRNA-L
SMILES[Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only