Benzyl chloromethyl ether, tech. 70%
Benzyl chloromethyl ether, tech. 70%
Benzyl chloromethyl ether, tech. 70%
Thermo Scientific Chemicals

Benzyl chloromethyl ether, tech. 70%

CAS: 3587-60-8 | C8H9ClO | 156.609 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
Catalog number H60207.AD
also known as H60207-AD
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
134.00
Save 20.35 (15%)
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
134.00
Save 20.35 (15%)
Each
Chemical Identifiers
CAS3587-60-8
IUPAC Name[(chloromethoxy)methyl]benzene
Molecular FormulaC8H9ClO
InChI KeyLADPCMZCENPFGV-UHFFFAOYSA-N
SMILESClCOCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>70.0%
Benzyl chloromethyl ether is employed in the introduction of benzoyloxy- and hydroxymethyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl chloromethyl ether is employed in the introduction of benzoyloxy- and hydroxymethyl groups.

Solubility
Soluble in water [2.7 g/L (25 C)]

Notes
Store at 2-8°C. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Dane A. Goff.; Ralph N. HarrisIII.; Jeffrey C. Bottaro.; Clifford D. Bedford. Cleavage of methoxymethyl ethers with boron trichloride. A convenient, versatile preparation of chloromethyl ether derivatives. J. Org. Chem. 1986, 51 (24),4711-4714.
  2. Arthur J. Hill.; DeWitt T. Keach. Some Ether-Substituted Derivatives Of Diethyl Malonate And Barbituric Acid.J. Am. Chem. Soc. 1926, 48 (1),257-262.