2-Chloromethyl-3,4-dimethoxypyridine hydrochloride, 97%, Thermo Scientific Chemicals
2-Chloromethyl-3,4-dimethoxypyridine hydrochloride, 97%, Thermo Scientific Chemicals
2-Chloromethyl-3,4-dimethoxypyridine hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloromethyl-3,4-dimethoxypyridine hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H60235.06
also known as H60235-06
5 g
Catalog number H60235.06
also known as H60235-06
Price (USD)
50.65
Online Exclusive
56.60
Save 5.95 (11%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
50.65
Online Exclusive
56.60
Save 5.95 (11%)
Each
Chemical Identifiers
CAS72830-09-2
IUPAC Name2-(chloromethyl)-3,4-dimethoxypyridine hydrochloride
Molecular FormulaC8H11Cl2NO2
InChI KeyYYRIKJFWBIEEDH-UHFFFAOYSA-N
SMILESCl.COC1=C(OC)C(CCl)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only