10-Bromodecanoic acid, 95%
10-Bromodecanoic acid, 95%
10-Bromodecanoic acid, 95%
Thermo Scientific Chemicals

10-Bromodecanoic acid, 95%

CAS: 50530-12-6 | C10H19BrO2 | 251.164 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H60292.06
also known as H60292-06
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.00
Save 6.35 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.00
Save 6.35 (14%)
Each
Chemical Identifiers
CAS50530-12-6
IUPAC Name10-bromodecanoic acid
Molecular FormulaC10H19BrO2
InChI KeyPGVRSPIEZYGOAD-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)>94.0%
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Partly miscible in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. F. L. M. Pattison.; J. B. Stothers.; R. G. Woolford. Anodic Syntheses Involving ι-Monohalocarboxylic Acids. J. Am. Chem. Soc. 1956, 78 (10), 2255-2259.
  2. Ali Dehshahri.; Reza K. Oskuee.; Wayne T. Shier.; Arash Hatefi.; Mohammad Ramezani. Gene transfer efficiency of high primary amine content, hydrophobic, alkyl-oligoamine derivatives of polyethylenimine. Biomaterials. 2009, 30 (25),4187-4194.