4-Bromotriphenylamine, 99%
4-Bromotriphenylamine, 99%
4-Bromotriphenylamine, 99%
4-Bromotriphenylamine, 99%
Thermo Scientific Chemicals

4-Bromotriphenylamine, 99%

CAS: 36809-26-4 | C18H14BrN | 324.221 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H60328.14
also known as H60328-14
Price (USD)
309.65
Online Exclusive
344.00
Save 34.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
309.65
Online Exclusive
344.00
Save 34.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS36809-26-4
IUPAC Name4-bromo-N,N-diphenylaniline
Molecular FormulaC18H14BrN
InChI KeySQTLUXJWUCHKMT-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only