Specifications
CAS325142-81-2
Chemical Name or Material2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester
Formula Weight294.03
IUPAC Name4,4,5,5-tetramethyl-2-(2,3,4,5,6-pentafluorophenyl)-1,3,2-dioxaborolane
InChI KeyFTJVUPKSBLYGSY-UHFFFAOYSA-N
View more