2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester, 96%
2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester, 96%
2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester, 96%
Thermo Scientific Chemicals

2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester, 96%

CAS: 325142-81-2 | C12H12BF5O2 | 294.028 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H60431.MD
also known as H60431-MD
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Specifications
CAS325142-81-2
Chemical Name or Material2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester
Formula Weight294.03
IUPAC Name4,4,5,5-tetramethyl-2-(2,3,4,5,6-pentafluorophenyl)-1,3,2-dioxaborolane
InChI KeyFTJVUPKSBLYGSY-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only