3,4,6-Tri-O-benzyl-D-glucal, 97%
3,4,6-Tri-O-benzyl-D-glucal, 97%
3,4,6-Tri-O-benzyl-D-glucal, 97%
Thermo Scientific Chemicals

3,4,6-Tri-O-benzyl-D-glucal, 97%

CAS: 55628-54-1 | C27H28O4 | 416.517 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H60437.03
also known as H60437-03
Price (USD)
51.80
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
51.80
Each
Specifications
Beilstein1691968
CAS55628-54-1
Chemical Name or Material3,4,6-Tri-O-benzyl-D-glucal
Formula Weight416.5
IUPAC Name(2R,3S,4R)-3,4-bis(phenylmethoxy)-2-(phenylmethoxymethyl)-3,4-dihydro-2H-pyran
View more
3,4,6-Tri-O-benzyl-D-glucal, is an important building block for both solid- and solution-phase synthesis of oligosaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,6-Tri-O-benzyl-D-glucal, is an important building block for both solid- and solution-phase synthesis of oligosaccharides.

Solubility
Insoluble in water, easily soluble in CHCl3

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. RW Scott.; CH Heathcock. An efficient synthesis of 3, 4, 6-tri-O-benzyl-2-C-methyl-d-glucal. Carbohydrate research. 1996291 (1), 205-208.
  2. V Bolitt.; C Mioskowski.; SG Lee. Direct preparation of 2-deoxy-D-glucopyranosides from glucals without Ferrier rearrangement. J. Org. Chem. 199055 (23), 5812-5813.