Oleanolic acid, 97%
Oleanolic acid, 97%
Oleanolic acid, 97%
Thermo Scientific Chemicals

Oleanolic acid, 97%

CAS: 508-02-1 | C30H48O3 | 456.711 g/mol
Have Questions?
Catalog NumberQuantity
H60445.MD
also known as H60445-MD
250 mg
Catalog number H60445.MD
also known as H60445-MD
Price (USD)
193.00
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
193.00
Each
Add to cart
Chemical Identifiers
CAS508-02-1
IUPAC Name(4aS,6aS,6bR,8aR,10S,12aR,12bR,14bS)-10-hydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylic acid
Molecular FormulaC30H48O3
InChI KeyMIJYXULNPSFWEK-GTOFXWBISA-N
SMILESCC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥96.0% (H-NMR)
FormCrystals or powder or crystalline powder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only