Methyl DL-lactate, 99%
Methyl DL-lactate, 99%
Methyl DL-lactate, 99%
Thermo Scientific Chemicals

Methyl DL-lactate, 99%

CAS: 547-64-8 | C4H8O3 | 104.105 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Catalog number H60455.AC
also known as H60455-AC
Price (USD)
217.65
Online Exclusive
242.00
Save 24.35 (10%)
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
217.65
Online Exclusive
242.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS547-64-8
IUPAC Namemethyl 2-hydroxypropanoate
Molecular FormulaC4H8O3
InChI KeyLPEKGGXMPWTOCB-UHFFFAOYNA-N
SMILESCOC(=O)C(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>98.5%
Methyl DL-lactate is used as a solvent for nitrocellulose, cellulase acetate, cellulase acetobutyrote and cellulase acetaprapionate. It is used in the manufacture of lacquers and dopes where it contributes high tolerance for diluents, good flaw and blush resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl DL-lactate is used as a solvent for nitrocellulose, cellulase acetate, cellulase acetobutyrote and cellulase acetaprapionate. It is used in the manufacture of lacquers and dopes where it contributes high tolerance for diluents, good flaw and blush resistance.

Solubility
Decomposes in water.

Notes
Moisture Sensitive. Store away from moisture and strong bases. Do not store together with acids. Store under dry inert gas. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Filip de Clippel; Michiel Dusselier; Ruben Van Rompaey; Pieter Vanelderen; Jan Dijkmans; Ekaterina Makshina; Lars Giebeler; Steffen Oswald; Gino V Baron; Joeri F M Denayer; Paolo P Pescarmona; Pierre A Jacobs; Bert F Sels. Fast and selective sugar conversion to alkyl lactate and lactic acid with bifunctional carbon-silica catalysts. Journal of the American Chemical Society.2012, 134 (24), 10089-10101.
  2. Debora Scuderi; Katia Le Barbu-Debus; A Zehnacker. The role of weak hydrogen bonds in chiral recognition. Physical Chemistry Chemical Physics.2011, 13 (40), 17916-17929.