5-Bromo-2,4-dimethoxybenzoic acid, 97%
5-Bromo-2,4-dimethoxybenzoic acid, 97%
5-Bromo-2,4-dimethoxybenzoic acid, 97%
Thermo Scientific Chemicals

5-Bromo-2,4-dimethoxybenzoic acid, 97%

CAS: 32246-20-1 | C9H9BrO4 | 261.071 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H60599.03
also known as H60599-03
Price (USD)
57.50
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
57.50
Each
Specifications
CAS32246-20-1
Chemical Name or Material5-Bromo-2,4-dimethoxybenzoic acid
Formula Weight261.07
IUPAC Name5-bromo-2,4-dimethoxybenzoic acid
InChI KeyWOPJFSQYDOHZMK-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only