3,5-Dibromophenol, 97%
3,5-Dibromophenol, 97%
3,5-Dibromophenol, 97%
Thermo Scientific Chemicals

3,5-Dibromophenol, 97%

CAS: 626-41-5 | C6H4Br2O | 251.905 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number H60642.09
also known as H60642-09
Price (USD)
185.65
Online Exclusive
206.00
Save 20.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
185.65
Online Exclusive
206.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS626-41-5
IUPAC Name3,5-dibromophenol
Molecular FormulaC6H4Br2O
InChI KeyPZFMWYNHJFZBPO-UHFFFAOYSA-N
SMILESOC1=CC(Br)=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to dark orange
Assay (unspecified)>96.0%
FormSolid
3,5-Dibromophenol is used as a pharmaceutical intermediate. It is also used to prepare aromatic polyesters and drugs for arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dibromophenol is used as a pharmaceutical intermediate. It is also used to prepare aromatic polyesters and drugs for arthritis.

Solubility
Sightly soluble in water. Soluble in methanol.

Notes
Incompatible with strong oxidizing agents. Light sensitive.
RUO – Research Use Only

General References:

  1. Bendig, P.; Vetter, W. UV-Induced Formation of Bromophenols from Polybrominated Diphenyl Ethers. Environ. Sci. Technol. 2013, 47 (8), 3665-3670.
  2. Shu, L.; Wang, P.; Radinov, R.; Dominique, R.; Wright, J.; Alabanza, L. M.; Dong, Y. Process Research on a Phenoxybutyric Acid LTB4 Receptor Antagonist. Efficient Kilogram-Scale Synthesis of a 3,5-Bisarylphenol Core. Org. Process Res. Dev. 2013, 17 (1), 114-119.
  3. Suna, S.; Canning, C. B.; Bhargava, K.; Sun, X.; Zhu, W.; Zhou, N.; Zhang, Y.; Zhou, K. Polybrominated diphenyl ethers with potent and broad spectrum antimicrobial activity from the marine sponge Dysidea. Bioorg. Med. Chem. Lett. 2015, 25 (10), 2181-2183.
  4. Kan,Y.; Zhu, Y.; Liu, Z.; Zhang, L.; Chen, J.; Cao, Y. Hydrophilic Conjugated Polymers with Large Bandgaps and Deep-Lying HOMO Levels as an Efficient Cathode Interlayer in Inverted Polymer Solar Cells. Macromol. Rapid Commun. 2015, 36 (15), 1393-1401.