It is a DuPhos and BPE ligands which are highly efficient privileged ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a DuPhos and BPE ligands which are highly efficient privileged ligands.
Solubility
Insoluble in water.
Notes
Air sensitive. Store at room temperature.
RUO – Research Use Only
Marie-Luis Schirmer.; Sven Adomeit.; Thomas Werner. First Base-Free Catalytic Wittig Reaction.Org. Lett. 2015, 17 (12),3078-3081.