3-(Diphenylphosphino)propylamine, 97+%
3-(Diphenylphosphino)propylamine, 97+%
3-(Diphenylphosphino)propylamine, 97+%
Thermo Scientific Chemicals

3-(Diphenylphosphino)propylamine, 97+%

CAS: 16605-03-1 | C15H18NP | 243.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H60672.MD
also known as H60672-MD
Price (USD)
52.60
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
52.60
Each
Chemical Identifiers
CAS16605-03-1
IUPAC Name(3-aminopropyl)diphenylphosphane
Molecular FormulaC15H18NP
InChI KeyDQZWMOWSTWWMPP-UHFFFAOYSA-N
SMILESNCCCP(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay by NMR>97.0%
Appearance (Color)Colorless to pale yellow
3-(Diphenylphosphino)propylamine is used as a paharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Diphenylphosphino)propylamine is used as a paharmaceutical.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, acids.
RUO – Research Use Only

General References:

  1. Martin E. Bluhm; Olaf Walter; Manfred Döring. Chromium imine and amine complexes as homogeneous catalysts for the trimerisation and polymerisation of ethylene. Journal of Organometallic Chemistry. 2005, 690 (3), 713-721.
  2. Martin Zeltner; Alexander Schätz; Max L. Heftia and Wendelin J. Stark. Magnetothermally responsive C/Co@PNIPAM-nanoparticles enable preparation of self-separating phase-switching palladium catalysts. J. Mater. Chem. 2005, 21 (9), 2991-2996.