4-Vinyl-1,3-dioxolan-2-one, 99%
4-Vinyl-1,3-dioxolan-2-one, 99%
4-Vinyl-1,3-dioxolan-2-one, 99%
4-Vinyl-1,3-dioxolan-2-one, 99%
4-Vinyl-1,3-dioxolan-2-one, 99%
Thermo Scientific Chemicals

4-Vinyl-1,3-dioxolan-2-one, 99%

CAS: 4427-96-7 | C5H6O3 | 114.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number H60822.30
also known as H60822-30
Price (USD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS4427-96-7
IUPAC Name(4S)-4-ethenyl-1,3-dioxolan-2-one
Molecular FormulaC5H6O3
InChI KeyBJWMSGRKJIOCNR-BYPYZUCNSA-N
SMILESC=C[C@H]1COC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>98.5%
It is used to synthesize functional polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to synthesize functional polymers.

Solubility
Insoluble in water.

Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

General References:

  1. John H. Clements. Reactive Applications of Cyclic Alkylene Carbonates. Ind. Eng. Chem. Res. 2003, 42 (4),663-674.
  2. Shang-Shi Zhang.; Jia-Qiang Wu.; Ye-Xing Lao.; Xu-Ge Liu.; Yao Liu.; Wen-Xin Lv.; Dong-Hang Tan.; Yao-Fu Zeng.; Honggen Wang. Mild Rhodium(III)-Catalyzed C-H Allylation with 4-Vinyl-1,3-dioxolan-2-ones: Direct and Stereoselective Synthesis of (E)-Allylic Alcohols. Org. Lett. 2014, 16 (24),6412-6415.