Formamidine hydrochloride, 98%
Formamidine hydrochloride, 98%
Formamidine hydrochloride, 98%
Thermo Scientific Chemicals

Formamidine hydrochloride, 98%

CAS: 6313-33-3 | CH4ClN2 | 79.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H60947.06
also known as H60947-06
Price (USD)
50.65
Online exclusive
56.40
Save 5.75
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
50.65
Online exclusive
56.40
Save 5.75
Each
Chemical Identifiers
CAS6313-33-3
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
5-Methyl-4,6-dihydroxypyrimidine was synthesized from formamidine hydrochloride. N-Formimino-n-glutamate was synthesized from formamidine hydrochloride. Formamidine hydrochloride was used in the synthesis of imidazoleglycerol phosphate (IGP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-4,6-dihydroxypyrimidine was synthesized from formamidine hydrochloride. N-Formimino-n-glutamate was synthesized from formamidine hydrochloride. Formamidine hydrochloride was used in the synthesis of imidazoleglycerol phosphate (IGP).

Solubility
Soluble in Ethanol.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water.
RUO – Research Use Only

General References:

  1. Shan Liu,; Qinghe Gao,; Xia Wu,; Jingjing Zhang,; Kerong Ding,; Anxin Wu. Formamidine hydrochloride as an amino surrogate: I2-catalyzed oxidative amidation of aryl methyl ketones leading to free (N-H) α-ketoamides. Org. Biomol. Chem. 2015, 13 issue , 2239-2242.
  2. H. M. Novais,; S. Steenken. Reactions of oxidizing radicals with 4,6-dihydroxypyrimidines as model compounds for uracil, thymine, and cytosine. J. Phys. Chem. 1977, 91 (2), 426-433.