7-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals
7-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals
7-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H61032.03
also known as H61032-03
1 g
Catalog number H61032.03
also known as H61032-03
Price (USD)
179.65
Special offer
Online exclusive
Ends: 28-Dec-2026
211.00
Save 31.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
179.65
Special offer
Online exclusive
Ends: 28-Dec-2026
211.00
Save 31.35
Each
Specifications
CAS120711-81-1
Chemical Name or Material7-Nitro-3,4-dihydro-2H-1,4-benzoxazine
Formula Weight180.16
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Sparingly soluble in water (0.76 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only