3,4-Dihydro-2H-1,4-benzothiazine, 97%
3,4-Dihydro-2H-1,4-benzothiazine, 97%
3,4-Dihydro-2H-1,4-benzothiazine, 97%
3,4-Dihydro-2H-1,4-benzothiazine, 97%
Thermo Scientific Chemicals

3,4-Dihydro-2H-1,4-benzothiazine, 97%

CAS: 3080-99-7 | C8H9NS | 151.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61045.03
also known as H61045-03
Price (USD)
64.65
Special offer
Online exclusive
Ends: 30-Jun-2026
75.80
Save 11.15 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
64.65
Special offer
Online exclusive
Ends: 30-Jun-2026
75.80
Save 11.15 (15%)
Each
Add to cart
Specifications
CAS3080-99-7
Chemical Name or Material3,4-Dihydro-2H-1,4-benzothiazine
Formula Weight151.23
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
3,4-Dihydro-2H-1,4-benzothiazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydro-2H-1,4-benzothiazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Violetta Cecchetti.; Arnaldo Fravolini.; Renata Fringuelli.; Giuseppe Mascellani.; Piergiuseppe Pagella.; Maurizio Palmioli.; Giorgio Segre.; Patrizia Terni. Quinolonecarboxylic acids. 2. Synthesis and antibacterial evaluation of 7-oxo-2,3 dihydro-7H-pyrido[1,2,3-de][1,4]benzothiazine-6-carboxylic acids. J. Med. Chem. 1987, 30 (3), 465-473.
  2. Raj Nandan Prasad. Potential antihypertensive agents. III. 3,4-Dihydro-2H-1,4-benzothiazine derivatives. J. Med. Chem. 1969, 12 (2), 290-294.