Specifications
CAS69373-37-1
Chemical Name or Material7-Nitro-3,4-dihydro-2H-1,4-benzothiazine
Formula Weight196.22
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more