2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine, 97%
2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine, 97%
2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine, 97%
2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine, 97%
Thermo Scientific Chemicals

2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine, 97%

CAS: 866089-28-3 | C10H13NO | 163.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H61058.03
also known as H61058-03
Price (USD)
225.65
Special offer
Online exclusive
Ends: 30-Jun-2026
265.00
Save 39.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
225.65
Special offer
Online exclusive
Ends: 30-Jun-2026
265.00
Save 39.35 (15%)
Each
Add to cart
Specifications
CAS866089-28-3
Chemical Name or Material2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine
Formula Weight163.22
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
It possess various biological activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It possess various biological activities.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
RUO – Research Use Only

General References:

  1. Domenico Albanese.; Dario Landini.; Vittoria Lupi.; Michele Penso. Straightforward Synthesis of 2-Substituted 3,4-Dihydro-2H-1,4-benzoxazines under Solid-Liquid Phase Transfer Catalysis Conditions.Ind. Eng. Chem. Res. 2003, 42 (4),680-686.
  2. Daiwang Xu.; Angèle Chiaroni.; Maurice-Bernard Fleury.; Martine Largeron. Electrochemically Induced Cascade Reaction for the Assembly of Libraries of Biologically Relevant 1,4-Benzoxazine Derivatives.J. Org. Chem. 2006, 71 (17),6374-6381.