7-Bromo-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals
7-Bromo-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals
7-Bromo-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals
7-Bromo-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Bromo-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H61071.03
also known as H61071-03
1 g
Catalog number H61071.03
also known as H61071-03
Price (USD)
271.65
Online Exclusive
302.00
Save 30.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
271.65
Online Exclusive
302.00
Save 30.35 (10%)
Each
Add to cart
Specifications
CAS105679-22-9
Chemical Name or Material7-Bromo-3,4-dihydro-2H-1,4-benzoxazine
Formula Weight214.06
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only