2-Chloro-5-nitroimidazole, 95%
2-Chloro-5-nitroimidazole, 95%
2-Chloro-5-nitroimidazole, 95%
Thermo Scientific Chemicals

2-Chloro-5-nitroimidazole, 95%

CAS: 57531-37-0 | C3H2ClN3O2 | 147.518 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61141.06
also known as H61141-06
Price (USD)
244.65
Special offer
Online exclusive
Ends: 30-Jun-2026
288.00
Save 43.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
244.65
Special offer
Online exclusive
Ends: 30-Jun-2026
288.00
Save 43.35 (15%)
Each
Chemical Identifiers
CAS57531-37-0
IUPAC Name2-chloro-5-nitro-1H-imidazole
Molecular FormulaC3H2ClN3O2
InChI KeyBOJZBRDIZUHTCE-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CN=C(Cl)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only