2-Fluoro-3-methylaniline, 97%
2-Fluoro-3-methylaniline, 97%
2-Fluoro-3-methylaniline, 97%
Thermo Scientific Chemicals

2-Fluoro-3-methylaniline, 97%

CAS: 1978-33-2 | C7H8FN | 125.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H61143.14
also known as H61143-14
Price (USD)
170.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
170.00
Each
Chemical Identifiers
CAS1978-33-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
Assay from Suppliers CofA≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only