4-Bromo-2-fluorophenylacetic acid, 98%
4-Bromo-2-fluorophenylacetic acid, 98%
4-Bromo-2-fluorophenylacetic acid, 98%
4-Bromo-2-fluorophenylacetic acid, 98%
Thermo Scientific Chemicals

4-Bromo-2-fluorophenylacetic acid, 98%

CAS: 114897-92-6 | C8H6BrFO2 | 233.036 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H61176.06
also known as H61176-06
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS114897-92-6
IUPAC Name2-(4-bromo-2-fluorophenyl)acetic acid
Molecular FormulaC8H6BrFO2
InChI KeyPNBIYFPZODYMOO-UHFFFAOYSA-N
SMILESOC(=O)CC1=C(F)C=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only