4-(2,5-Dimethyl-1-pyrrolyl)benzonitrile, 98%
4-(2,5-Dimethyl-1-pyrrolyl)benzonitrile, 98%
4-(2,5-Dimethyl-1-pyrrolyl)benzonitrile, 98%
Thermo Scientific Chemicals

4-(2,5-Dimethyl-1-pyrrolyl)benzonitrile, 98%

CAS: 119516-86-8 | C13H12N2 | 196.253 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61178.03
also known as H61178-03
Price (USD)
66.65
Online exclusive
74.20
Save 7.55
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
66.65
Online exclusive
74.20
Save 7.55
Each
Specifications
CAS119516-86-8
Chemical Name or Material4-(2,5-Dimethyl-1-pyrrolyl)benzonitrile
DOT InformationTransport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Formula Weight196.25
Health Hazard 1H302+H312+H332-H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only