4-Hydroxy-5-iodopyrimidine, 95%, Thermo Scientific Chemicals
4-Hydroxy-5-iodopyrimidine, 95%, Thermo Scientific Chemicals
4-Hydroxy-5-iodopyrimidine, 95%, Thermo Scientific Chemicals
4-Hydroxy-5-iodopyrimidine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxy-5-iodopyrimidine, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H61190.03
also known as H61190-03
1 g
Catalog number H61190.03
also known as H61190-03
Price (USD)
139.65
Online Exclusive
155.00
Save 15.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
139.65
Online Exclusive
155.00
Save 15.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS4349-07-9
IUPAC Name5-iodo-3,4-dihydropyrimidin-4-one
Molecular FormulaC4H3IN2O
InChI KeySMIGOGPUTQXOKF-UHFFFAOYSA-N
SMILESIC1=CN=CNC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
4-Hydroxy-5-iodopyrimidine is an intermediate used in pharmaceuticals and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-5-iodopyrimidine is an intermediate used in pharmaceuticals and chemical research.

Solubility
Soluble in water (67 g/L) (25°C).

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
Lech Skulski; and Piotr Wroczynski . Some Heteroaromatic Organomercurials, Their Syntheses and Reactions. Molecules. 2001, 6 (12), 927-958.