Trimethylsilanol, 95%
Trimethylsilanol, 95%
Trimethylsilanol, 95%
Thermo Scientific Chemicals

Trimethylsilanol, 95%

CAS: 1066-40-6 | C3H10OSi| 90.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H61276.06
also known as H61276-06
Price (USD)
60.65
Online Exclusive
67.00
Save 6.35 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
60.65
Online Exclusive
67.00
Save 6.35 (9%)
Each
Chemical Identifiers
CAS1066-40-6
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Trimethylsilanol used as a hydrophobic coat on silicate surfaces. Used as an as an antimicrobial agent.

Solubility
Soluble in water (1.00 mg/ml).

Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. F. O. Stark; O. K. Johannson; G. E. Vogel; R. G. Chaffee; R. M. Lacefield. Interactions between trialkylsilanes and E-Glass or Aerosil surfaces. Reactions of trimethylsilanol, trimethylchlorosilane, and hexamethyldisilazane. J. Phys. Chem. 1968, 72(8), 2750-2754.
  2. R. Grümping; D. Mikolajczak; A. V. Hirner. Determination of trimethylsilanol in the environment by LT-GC/ICP-OES and GC-MS. Fresenius' Journal of Analytical Chemistry. 1998, 361(2), 133-139.