2-Amino-5-methylpyrazine, 98%
2-Amino-5-methylpyrazine, 98%
2-Amino-5-methylpyrazine, 98%
Thermo Scientific Chemicals

2-Amino-5-methylpyrazine, 98%

CAS: 5521-58-4 | C5H7N3 | 109.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H61286.MD
also known as H61286-MD
Price (USD)
38.40
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
38.40
Each
Chemical Identifiers
CAS5521-58-4
IUPAC Name5-methylpyrazin-2-amine
Molecular FormulaC5H7N3
InChI KeyZNQOALAKPLGUPH-UHFFFAOYSA-N
SMILESCC1=CN=C(N)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.5% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only