2-Hydrazino-4-(trifluoromethyl)pyrimidine, 97%
2-Hydrazino-4-(trifluoromethyl)pyrimidine, 97%
2-Hydrazino-4-(trifluoromethyl)pyrimidine, 97%
Thermo Scientific Chemicals

2-Hydrazino-4-(trifluoromethyl)pyrimidine, 97%

CAS: 197305-97-8 | C5H5F3N4 | 178.118 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61287.06
also known as H61287-06
Price (USD)
188.65
Special offer
Online exclusive
Ends: 30-Jun-2026
222.00
Save 33.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
188.65
Special offer
Online exclusive
Ends: 30-Jun-2026
222.00
Save 33.35 (15%)
Each
Chemical Identifiers
CAS197305-97-8
IUPAC Name2-hydrazinyl-4-(trifluoromethyl)pyrimidine
Molecular FormulaC5H5F3N4
InChI KeyXQCAINVUBSAUBJ-UHFFFAOYSA-N
SMILESNNC1=NC=CC(=N1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>96.0%
2-Hydrazino-4-(trifluoromethyl)pyrimidine may be used as a derivatization reagent for the simultaneous determination of testosterone (T) and dihydrotestosterone (DHT) in murine tissue and serum. Synthesis of organohydrazide chelate complexes of rhenium and technetium. Preparation of 1-(4-methoxyphenyl)ethanone [(4-trifluoromethyl)pyrimidine-2-yl]hydrazone (NAS-75).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydrazino-4-(trifluoromethyl)pyrimidine may be used as a derivatization reagent for the simultaneous determination of testosterone (T) and dihydrotestosterone (DHT) in murine tissue and serum. Synthesis of organohydrazide chelate complexes of rhenium and technetium. Preparation of 1-(4-methoxyphenyl)ethanone [(4-trifluoromethyl)pyrimidine-2-yl]hydrazone (NAS-75).

Solubility
Soluble in aqueous acid.

Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Yan Weng; Fang Xie; Li Xu; Dmitri Zagorevski; David C. Spink; Xinxin Ding; Yan Weng; Fang Xie; Li Xu; Dmitri Zagorevski; David C. Spink; Xinxin Ding. Analysis of testosterone and dihydrotestosterone in mouse tissues by liquid chromatography-electrospray ionization-tandem mass spectrometry. Analytical Biochemistry. 2010, 402(2), 121-128.
  2. Hirsch-Kuchma M, et al. The synthesis and characterization of technetium and rhenium hydrazinopyrimidine chelate complexes. J. Chem. Soc., Dalton Trans. 1997, 183185-88.