2-Amino-5-fluoro-3-nitropyridine, 98%, Thermo Scientific Chemicals
2-Amino-5-fluoro-3-nitropyridine, 98%, Thermo Scientific Chemicals
2-Amino-5-fluoro-3-nitropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-5-fluoro-3-nitropyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H61293.06
also known as H61293-06
5 g
Catalog number H61293.06
also known as H61293-06
Price (USD)
391.65
Special offer
Online exclusive
Ends: 30-Jun-2026
460.00
Save 68.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
391.65
Special offer
Online exclusive
Ends: 30-Jun-2026
460.00
Save 68.35 (15%)
Each
Chemical Identifiers
CAS212268-12-7
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only