2-(Diphenylphosphino)benzaldehyde, 97%
2-(Diphenylphosphino)benzaldehyde, 97%
2-(Diphenylphosphino)benzaldehyde, 97%
Thermo Scientific Chemicals

2-(Diphenylphosphino)benzaldehyde, 97%

CAS: 50777-76-9 | C19H15OP | 290.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61306.06
also known as H61306-06
Price (USD)
249.65
Special offer
Online exclusive
Ends: 30-Jun-2026
294.00
Save 44.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
249.65
Special offer
Online exclusive
Ends: 30-Jun-2026
294.00
Save 44.35 (15%)
Each
Chemical Identifiers
CAS50777-76-9
IUPAC Name2-(diphenylphosphanyl)benzaldehyde
Molecular FormulaC19H15OP
InChI KeyDRCPJRZHAJMWOU-UHFFFAOYSA-N
SMILESO=CC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only