Methyl 3-(2-bromophenyl)propionate, 98%
Methyl 3-(2-bromophenyl)propionate, 98%
Methyl 3-(2-bromophenyl)propionate, 98%
Thermo Scientific Chemicals

Methyl 3-(2-bromophenyl)propionate, 98%

CAS: 66191-86-4 | C10H11BrO2 | 243.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H61342.14
also known as H61342-14
Price (USD)
344.65
Online Exclusive
383.00
Save 38.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
344.65
Online Exclusive
383.00
Save 38.35 (10%)
Each
Chemical Identifiers
CAS66191-86-4
IUPAC Namemethyl 3-(2-bromophenyl)propanoate
Molecular FormulaC10H11BrO2
InChI KeyNIULOVGPRAJIJM-UHFFFAOYSA-N
SMILESCOC(=O)CCC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only